Learn More
Alkali Scientific ASI™ Miniwell Tray, Teraski Format, 83x58mm, 10µL Working Volume, Polystyrene (PS), 100/Case, 72-Well, DNase, RNase & Pyrogen-Free
SDP
Catalog No. 503029074
Encompass_Preferred
Designed for precision and reliability, these Terasaki-style mini trays are ideal for serotyping, HLA applications, microlymphocytotoxicity, cell cloning studies, protein crystallization, and sequencing reactions. Each tray features a specially treated polystyrene surface that enhances reagent and sample mixing, even after freezing and thawing. This surface treatment minimizes antibody affinity, even at high concentrations, and reduces protein adsorption for improved assay performance. With exceptional optical clarity, these trays are compatible with inverted microscopes for detailed analysis. Large, raised alphanumeric markings ensure easy identification, while the wells—featuring an inverted, truncated cone design—offer a working volume of 10µL. For convenient storage and organization, high- and low-profile trays are stackable when used with the included lid, while S-plates can be stacked without a lid.
Catalog No. 50-302-9074 Supplier Alkali Scientific Supplier No. MWT072
Designed for precision and reliability, these Terasaki-style mini trays are ideal for serotyping, HLA applications, microlymphocytotoxicity, cell cloning studies, protein crystallization, and sequencing reactions. Each tray features a specially treated polystyrene surface that enhances reagent and sample mixing, even after freezing and thawing. This surface treatment minimizes antibody affinity, even at high concentrations, and reduces protein adsorption for improved assay performance. With exceptional optical clarity, these trays are compatible with inverted microscopes for detailed analysis. Large, raised alphanumeric markings ensure easy identification, while the wells—featuring an inverted, truncated cone design—offer a working volume of 10µL. For convenient storage and organization, high- and low-profile trays are stackable when used with the included lid, while S-plates can be stacked without a lid.