Learn More
Sigma Aldrich Fine Chemicals Biosciences Minimum Essential Medium Eagle With Earle's salts, L-glutamine, and non-essential amino acids, without sodium bicarbonate, powder, suitable for cell culture | 10L
Catalog No. NC0322773
Encompass
Minimum Essential Medium Eagle With Earle's salts, L-glutamine, and non-essential amino acids, without sodium bicarbonate, powder, suitable for cell culture | 10L
Catalog No. NC0322773 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M064310L
Minimum Essential Medium Eagle With Earle's salts, L-glutamine, and non-essential amino acids, without sodium bicarbonate, powder, suitable for cell culture | 10L