Learn More
ATCC Mino; Mantle Cell Lymphoma; Human (Homo sapiens)
Catalog No. 502383981
Encompass
Mino cells are lymphoblasts isolated from the peripheral blood of a 64-year-old, White male with mantle cell lymphoma. It can be used for immunology research.
Catalog No. 50-238-3981 Supplier ATCC Supplier No. CRL3000
Mino cells are lymphoblasts isolated from the peripheral blood of a 64-year-old, White male with mantle cell lymphoma. It can be used for immunology research.