Learn More
Research Products International Corp Minocycline Hydrochloride, 1 Gram
SDP
Catalog No. 50213517
Encompass_Preferred
Minocycline Hydrochloride (HCl) is a tetracycline antibiotic. Minocycline inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, preventing the attachment of aminoacyl-tRNA to the mRNA-ribosome complex. This interference results in the inhibition of protein synthesis and ultimately inhibits bacterial growth. 
Minocycline is effective against a wide range of bacteria, including both Gram-positive and Gram-negative bacteria. It is often used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic.
WARNING! This product can expose you to Minocycline HCl, a chemical which is known to the State of California to cause birth defects and reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-213-517 Supplier Research Products International Corp Supplier No. M920001.0
Minocycline Hydrochloride (HCl) is a tetracycline antibiotic. Minocycline inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, preventing the attachment of aminoacyl-tRNA to the mRNA-ribosome complex. This interference results in the inhibition of protein synthesis and ultimately inhibits bacterial growth. 
Minocycline is effective against a wide range of bacteria, including both Gram-positive and Gram-negative bacteria. It is often used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic.
WARNING! This product can expose you to Minocycline HCl, a chemical which is known to the State of California to cause birth defects and reproductive harm. For more information go to www.P65Warnings.ca.gov.