Learn More
Medchemexpress LLC Minocycline-d7 | 10118-90-8 | 464.52 | C23H20D7N3O7 | 500 UG
SDP
Catalog No. 5000284718
Encompass_Preferred
Minocycline-d7 is a deuterium-labeled (d7) isotopologue of minocycline supplied as a reference standard for analytical and biochemical applications. It is intended for use as an internal standard in mass spectrometry and for tracer studies where isotopic labeling provides clear mass separation from the native compound.
  • Deuterium-labeled (d7) isotopologue.
  • Suitable as an internal standard for mass spectrometry.
  • Useful for tracer and quantitative analytical studies.
  • Chemical formula C23H20D7N3O7 and molecular weight 464.52.
  • Available in a 500 UG package for analytical use.
Catalog No. 50-002-84718 Supplier Medchemexpress LLC Supplier No. HYW654013500UG
Minocycline-d7 is a deuterium-labeled (d7) isotopologue of minocycline supplied as a reference standard for analytical and biochemical applications. It is intended for use as an internal standard in mass spectrometry and for tracer studies where isotopic labeling provides clear mass separation from the native compound.
  • Deuterium-labeled (d7) isotopologue.
  • Suitable as an internal standard for mass spectrometry.
  • Useful for tracer and quantitative analytical studies.
  • Chemical formula C23H20D7N3O7 and molecular weight 464.52.
  • Available in a 500 UG package for analytical use.