Learn More
Alkali Scientific Minocycline Hydrochloride, 1 Gram
SDP
Catalog No. 5000661632
Encompass_Preferred
Minocycline Hydrochloride, a tetracycline-class antibiotic, is available in a 1g powder form for research use. Minocycline inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, making it effective against a wide range of gram-positive and gram-negative bacteria. In addition to its antibacterial properties, minocycline has been studied for its anti-inflammatory and neuroprotective effects. This compound is used in laboratory studies involving antibiotic resistance, protein synthesis inhibition, and inflammation research. The 1g amount is suitable for preparing concentrated solutions for microbiological and biochemical studies, particularly those focused on bacterial pathogens.
Catalog No. 50-006-61632 Supplier Alkali Scientific Supplier No. MIN001CP
Minocycline Hydrochloride, a tetracycline-class antibiotic, is available in a 1g powder form for research use. Minocycline inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, making it effective against a wide range of gram-positive and gram-negative bacteria. In addition to its antibacterial properties, minocycline has been studied for its anti-inflammatory and neuroprotective effects. This compound is used in laboratory studies involving antibiotic resistance, protein synthesis inhibition, and inflammation research. The 1g amount is suitable for preparing concentrated solutions for microbiological and biochemical studies, particularly those focused on bacterial pathogens.