Learn More
STEMCELL Technologies Minocycline (Hydrochloride Hydrate), Size: 100 mg
SDP
Supplier:  STEMCELL Technologies 74114
Encompass
Model neurodegenerative diseases in animal models with Minocycline. This semi-synthetic tetracycline with anti-inflammatory and neuroprotective properties (Garrido-Mesa et al. Br J Pharmacol, 2013) is used to reprogram extended pluripotent stem cells from humans and mice (Yang et al. Cell, 2017) and reduce the production of interleukin-8 in human epidermal keratinocytes (Ishikawa et al. Antimicrob Agents Chemother, 2009). Minocycline is also used in enhancing survival of neural stem cells in the rat model of Ischemia (Sakata et al. J Neurosci, 2012) and reducing bone and cartilage damage in rheumatoid arthritis (Greenwald et al. J Rheumatol, 1987). It also supports neuroprotective effects in animal models of various diseases, like Huntington’s disease (Chen et al. Nat Med, 2000), and Alzheimer’s disease (Choi et al. Neuropsychopharmacology, 2007). Minocycline is supplied as a hydrochloride salt with ≥98% purity; Molecular weight 493.9 g/mol. Visit STEMCELL.com for more information.
Catalog No. 50-197-6184