Learn More
Cayman Chemical Minocycline (hydrochloride hydrate), Purity: ≥98%, 100 mg
SDP
Catalog No. 502971544
Encompass_Preferred
A broad-spectrum tetracycline antibiotic; active against E. faecium in vitro (MIC = 16 mg/L); reduces the number of bacteria in the lung in a mouse model of multi-drug resistant A. baumannii-induced pneumonia at 20 mg/kg every 12 hours; reduces spinal cord lesion size and improves hindlimb function post-injury in a mouse model of acute spinal cord injury at an initial dose of 50 mg/kg and subsequent doses of 25 mg/kg per day for 5 days
Catalog No. 50-297-1544 Supplier Cayman Chemical Supplier No. 14454100 MG
A broad-spectrum tetracycline antibiotic; active against E. faecium in vitro (MIC = 16 mg/L); reduces the number of bacteria in the lung in a mouse model of multi-drug resistant A. baumannii-induced pneumonia at 20 mg/kg every 12 hours; reduces spinal cord lesion size and improves hindlimb function post-injury in a mouse model of acute spinal cord injury at an initial dose of 50 mg/kg and subsequent doses of 25 mg/kg per day for 5 days