Learn More
Medchemexpress LLC Minodronic acid | 180064-38-4 | 99.7% | 322.15 | 25 MG
SDP
Catalog No. 5000395047
Encompass_Preferred
Minodronic acid is a bisphosphonate that acts as an FPP synthase inhibitor and a P2X2/3 receptor antagonist. It has been shown to induce tumor cell apoptosis, inhibit cell growth, and suppress bone resorption, making it a valuable compound for research related to osteoporosis and various cancers.
  • FPP synthase inhibitor
  • Antagonist of P2X2/3 receptors
  • Induces tumor cell apoptosis
  • Inhibits cell growth
  • Suppresses bone resorption
  • Relevant for osteoporosis research
  • Relevant for cancer research
  • Appears as a white to off-white solid
Catalog No. 50-003-95047 Supplier Medchemexpress LLC Supplier No. 25MGHY16322
Add to Cart
Add to Cart
Minodronic acid is a bisphosphonate that acts as an FPP synthase inhibitor and a P2X2/3 receptor antagonist. It has been shown to induce tumor cell apoptosis, inhibit cell growth, and suppress bone resorption, making it a valuable compound for research related to osteoporosis and various cancers.
  • FPP synthase inhibitor
  • Antagonist of P2X2/3 receptors
  • Induces tumor cell apoptosis
  • Inhibits cell growth
  • Suppresses bone resorption
  • Relevant for osteoporosis research
  • Relevant for cancer research
  • Appears as a white to off-white solid