Learn More
Selleck Chemical LLC Minodronic acid S5057-25mg
SDP
Catalog No. 503112051
Encompass_Preferred
Minodronic acid (YM529 ONO-5920) is a third-generation nitrogen-cotaining bisphosphonate that is used for the treatment of osteoporosis Minodronic acid is an aminobisphosphonate that is a selective antagonist of purinergic P2X2/3 receptors involved in pain
Catalog No. 50-311-2051 Supplier Selleck Chemical LLC Supplier No. S505725MG
Add to Cart
Add to Cart
Minodronic acid (YM529 ONO-5920) is a third-generation nitrogen-cotaining bisphosphonate that is used for the treatment of osteoporosis Minodronic acid is an aminobisphosphonate that is a selective antagonist of purinergic P2X2/3 receptors involved in pain