Learn More
Precision Plus Vacuum Parts Inc Minor Repair Kit - Alcatel-Adixen 2004A, 2008A, 2012A, 2020A contains Buna o-rings
Catalog No. 501956204
Encompass_Preferred
Minor Repair Kits include coupling element, c-clips, o-rings, gaskets, washers, shaft seal, and other components for a light overhaul
Catalog No. 50-195-6204 Supplier Precision Plus Vacuum Parts Inc Supplier No. 52610B
Minor Repair Kits include coupling element, c-clips, o-rings, gaskets, washers, shaft seal, and other components for a light overhaul