Learn More
Precision Plus Vacuum Parts Inc Minor Repair Kit for Pfeiffer DUO1.6 and DUO3
Catalog No. 501956232
Encompass_Preferred
Minor Repair Kit includes sight glass gasket, o-rings, and shaft seals for minor pump maintenance.
Catalog No. 50-195-6232 Supplier Precision Plus Vacuum Parts Inc Supplier No. PKE01040T
Minor Repair Kit includes sight glass gasket, o-rings, and shaft seals for minor pump maintenance.