Learn More
Precision Plus Vacuum Parts Inc Minor Repair Kit for Pfeiffer DUO 30M
Catalog No. 501956246
Encompass_Preferred
Minor Repair Kit includes gasket, o-ring, and couplings for minor pump maintenance
Catalog No. 50-195-6246 Supplier Precision Plus Vacuum Parts Inc Supplier No. PKE30006T
Minor Repair Kit includes gasket, o-ring, and couplings for minor pump maintenance