Learn More
Precision Plus Vacuum Parts Inc Minor Repair Kit for Welch 2037, 2034, 2044
Catalog No. 501956265
Encompass_Preferred
Minor Repair Kit includes diaphragm, o-rings, and valve
Catalog No. 50-195-6265 Supplier Precision Plus Vacuum Parts Inc Supplier No. 2037K01
Minor Repair Kit includes diaphragm, o-rings, and valve