Learn More
Precision Plus Vacuum Parts Inc Minor Repair Kit for Welch 2062, 2064, 2067, MP1201Tp, MPC1201Tp, MP1801Zp, MPC1801Zp, MP2401Ep, MPC2401Ep
Catalog No. 501956262
Encompass_Preferred
Minor Repair Kit includes diaphragms, o-rings, and valve
Catalog No. 50-195-6262 Supplier Precision Plus Vacuum Parts Inc Supplier No. 402043
Minor Repair Kit includes diaphragms, o-rings, and valve