Learn More
HORIBA Instruments ABX MINOTROL 16 Hematology Tri-Level Control 12x2,.5mL
Catalog No. 22899104 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
12 x 2.5 mL
Catalog No. Quantity
22-899-104 12 x 2.5 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-899-104 Supplier HORIBA Instruments Supplier No. 5300100312
Add to Cart
Add to Cart
This item is not returnable. View return policy
ABX Minotrol 16 is a tri-level control intended for in vitro diagnostic use in monitoring the accuracy and precision on HORIBA ABX SAS hematology blood cell counters using HORIBA ABX SAS reagents.
Specifications
Quantity 12 x 2.5 mL