Learn More
American Radiolabeled Chemicals Inc MINOXIDIL 3-4-3H 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 071150UC
Encompass_Preferred
Minoxidil-piperyl-3,4-3H 50uCi, 30-60 Ci/mmol, ethanol, 1 mCi/ml, M.W. 209.25, Shipped in Dry Ice, Exclusive
Catalog No. 50-753-2629