Learn More
Medchemexpress LLC HY-B0112 500mg , Minoxidil CAS:38304-91-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0112500MG
Encompass_Preferred
HY-B0112 500mg Minoxidil CAS: 38304-91-5 Minoxidil (U10858) is an ATP-sensitive potassium (KATP) channel opener, a potent oral antihypertensive agent and a peripheral vasodilator that promotes vasodilation also affects hair growth. Minoxidil is also a potent inhibitor of soybean lipoxygenaseare with an IC50 of 20 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1117