Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minoxidil, Powder, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18604640 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-640 25 kg Amber Glass Bottle
18-604-635 25 g Amber Glass Bottle
18-604-636 100 g Amber Glass Bottle
18-604-637 500 g Amber Glass Bottle
18-604-638 1 kg Amber Glass Bottle
18-604-639 5 kg Amber Glass Bottle
18-604-633 1 g Amber Glass Bottle
18-604-634 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-604-640 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M113825KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

M1138, 38304-91-5, C9H15N5O

Spectrum™ Chemical Minoxidil, Powder, USP is an antihypertensive vasodilator used to help hair growth in the treatment of male pattern baldness. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 38304-91-5
Molecular Formula C9H15N5O
Molecular Weight (g/mol) 209.25
InChI Key ZIMGGGWCDYVHOY-UHFFFAOYSA-N
IUPAC Name 6-amino-2-imino-4-(piperidin-1-yl)-1,2-dihydropyrimidin-1-ol
SMILES NC1=CC(=NC(=N)N1O)N1CCCCC1

Specifications

CAS Max % 1
Ignition Residue 0.005
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 25 kg
Percent Purity 97 to 103%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Minoxidil, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.