Learn More
Medchemexpress LLC Minoxidil (Standard) | 38304-91-5 | 99.97% | C₉H₁₅N₅O | 250 MG
SDP
Catalog No. 5000358527
Encompass_Preferred
Minoxidil (Standard) is an analytical standard of Minoxidil, intended for research and analytical applications. It functions as an ATP-sensitive potassium (KATP) channel opener, a potent oral antihypertensive agent, and a peripheral vasodilator, promoting vasodilation and affecting hair growth. It also acts as a potent inhibitor of soybean lipoxygenase with an IC50 of 20 μM.
  • Analytical standard for research and analytical applications.
  • Functions as an ATP-sensitive potassium (KATP) channel opener.
  • Acts as a potent oral antihypertensive agent and peripheral vasodilator.
  • Promotes vasodilation and affects hair growth.
  • Potent inhibitor of soybean lipoxygenase with an IC50 of 20 μM.
  • Used in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS.
Catalog No. 50-003-58527 Supplier Medchemexpress LLC Supplier No. 250MGHYB0112R
Add to Cart
Add to Cart
Minoxidil (Standard) is an analytical standard of Minoxidil, intended for research and analytical applications. It functions as an ATP-sensitive potassium (KATP) channel opener, a potent oral antihypertensive agent, and a peripheral vasodilator, promoting vasodilation and affecting hair growth. It also acts as a potent inhibitor of soybean lipoxygenase with an IC50 of 20 μM.
  • Analytical standard for research and analytical applications.
  • Functions as an ATP-sensitive potassium (KATP) channel opener.
  • Acts as a potent oral antihypertensive agent and peripheral vasodilator.
  • Promotes vasodilation and affects hair growth.
  • Potent inhibitor of soybean lipoxygenase with an IC50 of 20 μM.
  • Used in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS.