Learn More
TARGETMOL CHEMICALS INC Minoxidil sulfate 50MG
SDP
Catalog No. 502877049
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Minoxidil sulfate (U-58838) is the active metabolite required to exert the vasodilatory and hair growing effects of minoxidil. Purity 99.93%
Catalog No. 50-287-7049 Supplier TARGETMOL CHEMICALS INC Supplier No. T813550MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Minoxidil sulfate (U-58838) is the active metabolite required to exert the vasodilatory and hair growing effects of minoxidil. Purity 99.93%