Learn More
Cayman Chemical Minoxidil Sulfate, Purity: ≥85%, CAS: 83701-22-8, 5 mg
SDP
Catalog No. 502911910
Encompass_Preferred
A hair follicle stimulator and an active metabolite of minoxidil; binds to KATP channels in a biphasic manner in COS-7 cells expressing the human channel (IC50s = 0.62 and 0.96 µM); induces vasodilation in isolated rabbit mesenteric artery at 5 µM
Catalog No. 50-291-1910 Supplier Cayman Chemical Supplier No. 360845 MG
A hair follicle stimulator and an active metabolite of minoxidil; binds to KATP channels in a biphasic manner in COS-7 cells expressing the human channel (IC50s = 0.62 and 0.96 µM); induces vasodilation in isolated rabbit mesenteric artery at 5 µM