Learn More
Medchemexpress LLC Minoxidil sulfate | 83701-22-8 | 98.0% | 1 ML

Supplier: Medchemexpress LLC HYB144510MM/1ML
Minoxidil sulfate is a potent and ATP-sensitive K+ channel opener and the sulfated metabolite of minoxidil. It functions as a vasodilator, promoting hair growth in vivo. It significantly reduces the permeability of the blood-brain tumor barrier (BTB) in a time-dependent manner and increases RhoA activation, inducing transient PKB phosphorylation at Ser-473 in brain tumor tissue.
- Potent and ATP-sensitive K+ channel opener.
- Sulfated metabolite of minoxidil.
- Considered a vasodilator that promotes hair growth in vivo.
- Significantly reduces the permeability of the blood-brain tumor barrier (BTB) in a time-dependent manner.
- Increases RhoA activation and induces transient PKB phosphorylation at Ser-473 in brain tumor tissue.
- Molecular weight is 289.31.
- Formula is C9H15N5O4S.
- Appears as a white to off-white solid.
- Soluble in DMSO at 100 mg/mL.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.