Learn More
Medchemexpress LLC Minoxidil sulfate | 83701-22-8 | 98.0% | 100 MG
SDP
Catalog No. 5000412610
Encompass_Preferred
Minoxidil sulfate is a potent and ATP-sensitive K+ channel opener, acting as the sulfated metabolite of minoxidil. It is considered a vasodilator that promotes hair growth in vivo. This product is for research use only and not sold to patients.
  • Potent and ATP-sensitive K+ channel opener.
  • Sulfated metabolite of minoxidil.
  • Considered a vasodilator to promote hair growth in vivo.
  • Reduces the permeability of blood-brain tumor barrier (BTB) in RBMEC monolayers.
  • Increases RhoA activation and induces transient PKB phosphorylation at Ser-473 in brain tumor tissue.
Catalog No. 50-004-12610 Supplier Medchemexpress LLC Supplier No. HYB1445100MG
Minoxidil sulfate is a potent and ATP-sensitive K+ channel opener, acting as the sulfated metabolite of minoxidil. It is considered a vasodilator that promotes hair growth in vivo. This product is for research use only and not sold to patients.
  • Potent and ATP-sensitive K+ channel opener.
  • Sulfated metabolite of minoxidil.
  • Considered a vasodilator to promote hair growth in vivo.
  • Reduces the permeability of blood-brain tumor barrier (BTB) in RBMEC monolayers.
  • Increases RhoA activation and induces transient PKB phosphorylation at Ser-473 in brain tumor tissue.