Learn More
Medchemexpress LLC Minoxidil sulfate | 83701-22-8 | 98.0% | 25 MG
SDP
Catalog No. 5000413903
Encompass_Preferred
Minoxidil sulfate is a potent and ATP-sensitive K+ channel opener. It is the sulfated metabolite of minoxidil and is considered a vasodilator that promotes hair growth in vivo.
  • Potent and ATP-sensitive K+ channel opener
  • Sulfated metabolite of minoxidil
  • Considered a vasodilator to promote hair growth in vivo
Catalog No. 50-004-13903 Supplier Medchemexpress LLC Supplier No. 25MGHYB1445
Minoxidil sulfate is a potent and ATP-sensitive K+ channel opener. It is the sulfated metabolite of minoxidil and is considered a vasodilator that promotes hair growth in vivo.
  • Potent and ATP-sensitive K+ channel opener
  • Sulfated metabolite of minoxidil
  • Considered a vasodilator to promote hair growth in vivo