Learn More
Selleck Chemical LLC Minoxidil sulphate S5228-1g
SDP
Catalog No. 503111889
Encompass_Preferred
Minoxidil (U-58838) is a potent direct-acting peripheral vasodilator that reduces peripheral resistance and produces a fall in blood pressure
Catalog No. 50-311-1889 Supplier Selleck Chemical LLC Supplier No. S52281G
Minoxidil (U-58838) is a potent direct-acting peripheral vasodilator that reduces peripheral resistance and produces a fall in blood pressure