Learn More
Chemglass Life Sciences Thermometer, -10 to 150°C In 1°C Subdivision, Overall Length = 152mm
SDP
Catalog No. 50872603
Encompass_Preferred
Thermometer, -10 to 150¿C In 1¿C Subdivision, Overall Length = 152mm. Bulb immersion, mineral spirit filled, thermometer with engraved stem. Stem OD is 5.5mm to 6.0mm with overall length as listed. Each thermometer is supplied with a #106 Viton O-ring for suspension of thermometer in open distillation systems.
Catalog No. 50-872-603 Supplier Chemglass Life Sciences Supplier No. MW110N01
Thermometer, -10 to 150¿C In 1¿C Subdivision, Overall Length = 152mm. Bulb immersion, mineral spirit filled, thermometer with engraved stem. Stem OD is 5.5mm to 6.0mm with overall length as listed. Each thermometer is supplied with a #106 Viton O-ring for suspension of thermometer in open distillation systems.