Learn More
Chemglass Life Sciences Thermometer, -10 to 250°C In 2°C Subdivision, Overall Length = 225mm
SDP
Catalog No. 50872604
Encompass_Preferred
Thermometer, -10 to 250¿C In 2¿C Subdivision, Overall Length = 225mm. Bulb immersion, mineral spirit filled, thermometer with engraved stem. Stem OD is 5.5mm to 6.0mm with overall length as listed. Each thermometer is supplied with a #106 Viton O-ring for suspension of thermometer in open distillation systems.
Catalog No. 50-872-604 Supplier Chemglass Life Sciences Supplier No. MW110N02
Thermometer, -10 to 250¿C In 2¿C Subdivision, Overall Length = 225mm. Bulb immersion, mineral spirit filled, thermometer with engraved stem. Stem OD is 5.5mm to 6.0mm with overall length as listed. Each thermometer is supplied with a #106 Viton O-ring for suspension of thermometer in open distillation systems.