Learn More
MSC Vermont Gage Class ZZ Minus Plug Gage, 0.3490 in. Diameter, 52100 Tool Steel, 1 ea.
Catalog No. 5000510284
Encompass
This Class ZZ minus plug and pin gage is designed for precise measurement of hole sizes, setting micrometers, and use in NoGo inspection applications. Manufactured from 52100 tool steel, this gage is heat treated to a hardness of 60 to 65 Rc for durability and wear resistance.
  • Diameter of 0.3490 in.
  • Overall length of 2 in.
  • Tolerance of -0.0002 in./+0.0000 in.
  • Centerless lapped finish of 10 microin. or better
  • Black oxide coating
  • Size electro etched on each piece
  • NIST traceable with included certificate of accuracy
Catalog No. 50-005-10284 Supplier MSC Supplier No. 87888053
This Class ZZ minus plug and pin gage is designed for precise measurement of hole sizes, setting micrometers, and use in NoGo inspection applications. Manufactured from 52100 tool steel, this gage is heat treated to a hardness of 60 to 65 Rc for durability and wear resistance.
  • Diameter of 0.3490 in.
  • Overall length of 2 in.
  • Tolerance of -0.0002 in./+0.0000 in.
  • Centerless lapped finish of 10 microin. or better
  • Black oxide coating
  • Size electro etched on each piece
  • NIST traceable with included certificate of accuracy