Learn More
MSC Vermont Gage Class ZZ Minus Plug Gage, 0.2845 in. Diameter, Steel, 2 in. Length, 1 ea.
Catalog No. 5000514754
Encompass
This Class ZZ minus plug gage is a high-precision measurement tool designed for inspecting, setting micrometers, and checking verniers. It features a precise diameter of 0.2845 in. and an overall length of 2 in., engineered for reliable performance in demanding quality control and metalworking environments.
  • Hardness of 60 to 62 Rc
  • Finish of 10 microinch or better
  • Roundness within 0.0001 in. (0.0025 mm)
  • Tolerance of -0.0002 in./+0.0000 in.
  • Includes a certificate of accuracy with NIST traceability
  • Centerless lapped for enhanced surface quality
  • Marked with size and direction of tolerance
Catalog No. 50-005-14754 Supplier MSC Supplier No. 59324475
This Class ZZ minus plug gage is a high-precision measurement tool designed for inspecting, setting micrometers, and checking verniers. It features a precise diameter of 0.2845 in. and an overall length of 2 in., engineered for reliable performance in demanding quality control and metalworking environments.
  • Hardness of 60 to 62 Rc
  • Finish of 10 microinch or better
  • Roundness within 0.0001 in. (0.0025 mm)
  • Tolerance of -0.0002 in./+0.0000 in.
  • Includes a certificate of accuracy with NIST traceability
  • Centerless lapped for enhanced surface quality
  • Marked with size and direction of tolerance