Learn More
MSC Vermont Gage Pin Gage, 0.4450 in. Diameter, Class ZZ, 2 in. Length, 1 ea.
Catalog No. 5000547966
Encompass
This Class ZZ minus plug and pin gage is a precision-machined cylindrical tool designed for accurate measuring and inspecting applications. Constructed from high-quality 52100 tool steel, this gage is suitable for setting micrometers, checking verniers, and performing No-Go inspections on various measuring devices.
  • Features a 0.4450 in. diameter and 2 in. overall length
  • Maintains a tolerance class of ZZ with a minus direction of 0.0002 in.
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from durable 52100 tool steel
  • Features a black oxide coating for enhanced wear resistance
  • Designed as a reversible pin gage for versatile use
Catalog No. 50-005-47966 Supplier MSC Supplier No. 87354965
This Class ZZ minus plug and pin gage is a precision-machined cylindrical tool designed for accurate measuring and inspecting applications. Constructed from high-quality 52100 tool steel, this gage is suitable for setting micrometers, checking verniers, and performing No-Go inspections on various measuring devices.
  • Features a 0.4450 in. diameter and 2 in. overall length
  • Maintains a tolerance class of ZZ with a minus direction of 0.0002 in.
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from durable 52100 tool steel
  • Features a black oxide coating for enhanced wear resistance
  • Designed as a reversible pin gage for versatile use