Learn More
MSC Vermont Gage Class ZZ No-Go Pin Gage, Steel, 0.2820 in. Diameter, 2 in. Length, 1 ea
Catalog No. 5000511229
Encompass
This Class ZZ No-Go pin gage provides a precise measurement of 0.2820 in. for industrial quality control applications. Manufactured from durable tool steel, this cylindrical gage is ideal for calibrating measuring devices or performing No-Go inspection tasks. It features a black oxide finish and is supplied with a certificate of accuracy.
  • Diameter of 0.2820 in.
  • Overall length of 2 in.
  • Hardness of 60/65 Rc
  • Surface finish of 10 microinch or better
  • Meets ISO 17025:2005 standards
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-11229 Supplier MSC Supplier No. 50736735
This Class ZZ No-Go pin gage provides a precise measurement of 0.2820 in. for industrial quality control applications. Manufactured from durable tool steel, this cylindrical gage is ideal for calibrating measuring devices or performing No-Go inspection tasks. It features a black oxide finish and is supplied with a certificate of accuracy.
  • Diameter of 0.2820 in.
  • Overall length of 2 in.
  • Hardness of 60/65 Rc
  • Surface finish of 10 microinch or better
  • Meets ISO 17025:2005 standards
  • Includes certificate of accuracy with NIST traceability