Learn More
Invent Biotechnologies Inc Minute Single Nucleus Isolation Kit for Tissues/Cells, Catalog Number SN-047, Processing Time ~30 min, Minimum Tissue Input 1 mg, Recommended Tissue Input 15-20 mg, Minimum Cell Input 200,000 cells, Recommended Cell Input 4-5 million cells, 20 preps
Catalog No. NC1755003
Encompass
The Minute Single Nucleus Isolation Kit is designed for the efficient isolation and purification of single nuclei from various tissue and cell types. This kit overcomes common challenges in traditional nuclei isolation, such as aggregation and doublet formation, by providing a simple, rapid, and highly effective protocol. It is ideal for applications that require high-quality, well-separated single nuclei, including FACS analysis, single nucleus RNA-seq, immunofluorescence staining, and cell cycle analysis.
  • Enhanced single nuclei yield to minimize aggregation and doublet formation.
  • Rapid processing time of approximately 30 minutes for nuclei isolation.
  • Broad compatibility with fresh or frozen tissues and cultured cells.
  • Low input requirement that works with 1-30 mg of tissue.
  • Suitable for diverse downstream applications such as RNA-seq and ATAC-seq.
Catalog No. NC1755003 Supplier Invent Biotechnologies Inc Supplier No. SN047
The Minute Single Nucleus Isolation Kit is designed for the efficient isolation and purification of single nuclei from various tissue and cell types. This kit overcomes common challenges in traditional nuclei isolation, such as aggregation and doublet formation, by providing a simple, rapid, and highly effective protocol. It is ideal for applications that require high-quality, well-separated single nuclei, including FACS analysis, single nucleus RNA-seq, immunofluorescence staining, and cell cycle analysis.
  • Enhanced single nuclei yield to minimize aggregation and doublet formation.
  • Rapid processing time of approximately 30 minutes for nuclei isolation.
  • Broad compatibility with fresh or frozen tissues and cultured cells.
  • Low input requirement that works with 1-30 mg of tissue.
  • Suitable for diverse downstream applications such as RNA-seq and ATAC-seq.