Learn More
Minuteman™ CRV Vacuum Accessories
SDP
Catalog No. 18999285 Shop All Minuteman International, Inc. Products
Click to view available options
Product Type:
Filter Cloth Bag Assembly
ULPA Filter Assembly
Quantity:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
18-999-285 ULPA Filter Assembly Each
18-999-283 Filter Cloth Bag Assembly Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-285 Supplier Minuteman™ Supplier No. 110039
Filters for use with CRV Cleanroom Vacuums
Specifications
For Use With (Application) Class 1 through Class 100,000 Cleanrooms Vacuuming
Product Type ULPA Filter Assembly
Quantity Each
For Use With (Equipment) CRV and RFI/EMI Vacuum Filtration Systems