Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse MIP-1 gamma (CCL9) ELISA Kit

Catalog No. EMCCL9X5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EMCCL9X5 5 x 96 Tests
EMCCL9 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EMCCL9X5 Supplier Invitrogen™ Supplier No. EMCCL9X5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

The Mouse MIP-1 gamma (CCL9) ELISA quantitates Ms MIP-1γ in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms MIP-1γ. Principle of the method The Mouse MIP-1γ solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

MIP-1gamma is a CC chemokine found in murine blood and a wide variety of murine tissues, with no known human homolog. MIP-1gamma signals through the CCR1 receptor. MIP-1gamma chemoattracts neutrophils, and also inhibits colony formation of bone marrow myeloid immature progenitors. MIP-1gamma contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51670
Assay Range 1.02-250 pg/mL
Assay Sensitivity 1 pg/mL
Calibration Range 1 pg/mL to 250 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Chemokines & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP- 5, M-CSF, MIG, MIP-1 alpha, MIP-1 delta, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, PSelectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF.
Sufficient For 5 x 96 assays
Target Species Mouse
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias C-C motif chemokine 9; CCF18; Ccl9; CCL9(29-101); CCL9(30-101); CCL9(31-101); chemokine (C-C motif) ligand 9; chemokine CCL9; macrophage inflammatory protein 1-gamma; Macrophage inflammatory protein-related protein 2; MIP-1 gamma; MIP-1-gamma; Mrp2; MRP-2; RP23-320E6.3; Scya10; Scya9; small inducible cytokine A10; small inducible cytokine A9; Small-inducible cytokine A9C-C motif chemokine 9; CCF18; Ccl9; CCL9(29-101); CCL9(30-101); CCL9(31-101); chemokine (C-C motif) ligand 9; chemokine CCL9; macrophage inflammatory protein 1-gamma; Macrophage inflammatory protein-related protein 2; MIP-1 gamma; MIP-1-gamma; Mrp2; MRP-2; RP23-320E6.3; Scya10; Scya9; small inducible cytokine A10; small inducible cytokine A9; Small-inducible cytokine A9; CCF18; MRP-2; Scya10; Scya9
Gene ID (Entrez) 20308
Gene Symbol Ccl9
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Inflammation
Sample Volume Plasma 2.5 μL, Serum 2.5 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym RP23-320E6.3, CCF18, MRP-2, Scya10, Scya9
Target MIP-1 gamma
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.