Learn More
Medchemexpress LLC MIP-1095 (TFA) | 99.8% | 678.35 | 25 MG
SDP
Catalog No. 5000438426
Encompass_Preferred
MIP-1095 TFA (RPS-001 TFA) is a potent inhibitor of PSMA, demonstrating good biodistribution and efficient targeting of tumor lesions. It is utilized as an imaging probe to identify and monitor tumor progression, particularly when isotopically labeled with 131I. This labeling results in 131I-MIP-1095 showing higher renal uptake in mice.
  • Potent inhibitor of PSMA
  • Good biodistribution
  • Efficient targeting of tumor lesions
  • Can be isotopically labeled as an imaging probe
  • Higher renal uptake in mice when labeled with 131I
  • Used as an imaging probe to indicate tumor progression
Catalog No. 50-004-38426 Supplier Medchemexpress LLC Supplier No. 25MGHY123733A
MIP-1095 TFA (RPS-001 TFA) is a potent inhibitor of PSMA, demonstrating good biodistribution and efficient targeting of tumor lesions. It is utilized as an imaging probe to identify and monitor tumor progression, particularly when isotopically labeled with 131I. This labeling results in 131I-MIP-1095 showing higher renal uptake in mice.
  • Potent inhibitor of PSMA
  • Good biodistribution
  • Efficient targeting of tumor lesions
  • Can be isotopically labeled as an imaging probe
  • Higher renal uptake in mice when labeled with 131I
  • Used as an imaging probe to indicate tumor progression