Learn More
Medchemexpress LLC MIP-1095 trifluoroacetate | 00-00-0 | MFCD09802015 | 99.8% | 678.35 g/mol | C21H26F3IN4O10 | 5 MG
SDP
Catalog No. 5000264528
Encompass_Preferred
MIP-1095 trifluoroacetate is a small-molecule inhibitor of prostate-specific membrane antigen used for research and imaging probe development. The compound is supplied as a high-purity trifluoroacetate salt, suitable for radiolabeling and preclinical assays.
  • Potent inhibitor of prostate-specific membrane antigen (PSMA).
  • Provided as trifluoroacetate (TFA) salt for stability.
  • High purity suitable for research applications.
  • Compatible with radiolabeling for imaging probe development.
  • Soluble in DMSO for assay preparation.
  • Available in small-mass quantities for preclinical studies.
Catalog No. 50-002-64528 Supplier Medchemexpress LLC Supplier No. HY123733A5MG
MIP-1095 trifluoroacetate is a small-molecule inhibitor of prostate-specific membrane antigen used for research and imaging probe development. The compound is supplied as a high-purity trifluoroacetate salt, suitable for radiolabeling and preclinical assays.
  • Potent inhibitor of prostate-specific membrane antigen (PSMA).
  • Provided as trifluoroacetate (TFA) salt for stability.
  • High purity suitable for research applications.
  • Compatible with radiolabeling for imaging probe development.
  • Soluble in DMSO for assay preparation.
  • Available in small-mass quantities for preclinical studies.