Learn More
American Radiolabeled Chemicals Inc MIP-1A-125I 25UC
SDP
Catalog No. 50901035
Encompass
MIP-1a-125I (human, recombinant) 25uCi in buffered solution (pH 4-4.2), 0.025 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~8000
Catalog No. 50-901-035 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 014925UC
Add to Cart
Add to Cart
Request a Quote
MIP-1a-125I (human, recombinant) 25uCi in buffered solution (pH 4-4.2), 0.025 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~8000