Learn More
Medchemexpress LLC HY-P7258 10ug Medchemexpress, MIP-2/CXCL2, Mouse CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP725810UG
Encompass_Preferred
Medchemexpress, HY-P7258 10ug MIP-2/CXCL2, Mouse CAS: Recombinant Mouse Macrophage Inflammatory Protein-2/CXCL2 is a member of the CXC chemokine family, and can recruit neutrophils to infection sites. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4370