Learn More
Medchemexpress LLC HY-P7262 10ug Medchemexpress, MIP-3-alpha/CCL20, Human CAS: Purity:>98%
SDP
Catalog No. 501934371
Encompass_Preferred
Medchemexpress, HY-P7262 10ug MIP-3-alpha/CCL20, Human CAS: Recombinant Human MIP-3 alpha/CCL20 is a C-C motif chemokine ligand, and has CCR6 as its sole receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4371 Supplier Medchemexpress LLC Supplier No. HYP726210UG
Medchemexpress, HY-P7262 10ug MIP-3-alpha/CCL20, Human CAS: Recombinant Human MIP-3 alpha/CCL20 is a C-C motif chemokine ligand, and has CCR6 as its sole receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.