Learn More
Medchemexpress LLC HY-P7263 10ug Medchemexpress, MIP-3-alpha/CCL20, Mouse CAS: Purity:>98%
SDP
Catalog No. 502083136
Encompass_Preferred
Medchemexpress, HY-P7263 10ug MIP-3-alpha/CCL20, Mouse CAS: Recombinant Mouse Macrophage Inflammatory Protein-3 alpha/CCL20 is a C-C motif chemokine ligand, and has CCR6 as its sole receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3136 Supplier Medchemexpress LLC Supplier No. HYP726310UG
Medchemexpress, HY-P7263 10ug MIP-3-alpha/CCL20, Mouse CAS: Recombinant Mouse Macrophage Inflammatory Protein-3 alpha/CCL20 is a C-C motif chemokine ligand, and has CCR6 as its sole receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.