Learn More
Medchemexpress LLC HY-P7265 10ug Medchemexpress, MIP-4/CCL18, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP726510UG
Encompass_Preferred
Medchemexpress, HY-P7265 10ug MIP-4/CCL18, Human CAS: Recombinant Human MIP-4/CCL18 is a CC chemokine ligand, acts as a neutral CCR3 antagonist, and is involved in inflammatory response. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3137