Learn More
Medchemexpress LLC HY-P7266 10ug Medchemexpress, MIP-5/CCL15, Human CAS: Purity:>98%
SDP
Catalog No. 502083138
Encompass_Preferred
Medchemexpress, HY-P7266 10ug MIP-5/CCL15, Human CAS: Recombinant Human MIP-5/CCL15 is a chemokine with a strong chemotactic activity for myeloid cells such as dendritic cells, monocytes, neutrophils and some T-lymphocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3138 Supplier Medchemexpress LLC Supplier No. HYP726610UG
Medchemexpress, HY-P7266 10ug MIP-5/CCL15, Human CAS: Recombinant Human MIP-5/CCL15 is a chemokine with a strong chemotactic activity for myeloid cells such as dendritic cells, monocytes, neutrophils and some T-lymphocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.