Learn More
Zymo Research Corporation MIP Buffer, 20 ml
SDP
Catalog No. 502037135
Encompass_Preferred
The MIP Buffer provides optimal conditions for the antibody/Protein A/DNA complex formation. Highlights: Sold separately or as part of the Methylated-DNA IP Kit (D5101). Storage temperature: 0°C to room temperature.
Catalog No. 50-203-7135 Supplier Zymo Research Corporation Supplier No. D5101320
The MIP Buffer provides optimal conditions for the antibody/Protein A/DNA complex formation. Highlights: Sold separately or as part of the Methylated-DNA IP Kit (D5101). Storage temperature: 0°C to room temperature.