Learn More
Medchemexpress LLC Mipomersen (sodium) | 629167-92-6 | 97.5% | 7179.15 | 50 MG
SDP
Catalog No. 5000344890
Encompass_Preferred
Mipomersen sodium (ISIS 301012) is an antisense oligonucleotide inhibitor of apolipoprotein B (apoB). It has an anti-HCV effect and reduces the infectivity of HCV. Mipomersen sodium can be used for the research of homozygous familial hypercholesterolemia (HoFH). It predominantly distributes to the liver and decreases the production of apoB-100, reducing plasma LDL-cholesterol and apoB-100 concentrations.
  • Antisense oligonucleotide inhibitor of apolipoprotein B.
  • Anti-HCV effect and reduces the infectivity of HCV.
  • Can be used for the research of homozygous familial hypercholesterolemia (HoFH).
  • Predominantly distributes to the liver.
  • Decreases the production of apoB-100.
  • Reduces plasma LDL-cholesterol and apoB-100 concentrations.
Catalog No. 50-003-44890 Supplier Medchemexpress LLC Supplier No. HY1087641MG
Mipomersen sodium (ISIS 301012) is an antisense oligonucleotide inhibitor of apolipoprotein B (apoB). It has an anti-HCV effect and reduces the infectivity of HCV. Mipomersen sodium can be used for the research of homozygous familial hypercholesterolemia (HoFH). It predominantly distributes to the liver and decreases the production of apoB-100, reducing plasma LDL-cholesterol and apoB-100 concentrations.
  • Antisense oligonucleotide inhibitor of apolipoprotein B.
  • Anti-HCV effect and reduces the infectivity of HCV.
  • Can be used for the research of homozygous familial hypercholesterolemia (HoFH).
  • Predominantly distributes to the liver.
  • Decreases the production of apoB-100.
  • Reduces plasma LDL-cholesterol and apoB-100 concentrations.