Learn More
Medchemexpress LLC HY-13930 5mg Medchemexpress, Miquelianin CAS:22688-79-5 Purity:>98%
SDP
Catalog No. 502081533
Encompass_Preferred
Medchemexpress, HY-13930 5mg Miquelianin CAS:22688-79-5 Miquelianin (Quercetin 3-O-glucuronide) is a metabolite of quercetin and a type of natural flavonoid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1533 Supplier Medchemexpress LLC Supplier No. HY139305MG
Medchemexpress, HY-13930 5mg Miquelianin CAS:22688-79-5 Miquelianin (Quercetin 3-O-glucuronide) is a metabolite of quercetin and a type of natural flavonoid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.