Learn More
BIOHIPPO miR-375-3p-GFP-Puro
SDP
Catalog No. 5000330746
Encompass
miR-375-3p-GFP-Puro lentiviral vector expressing microRNA 375 with GFP reporter and Puromycin selection for studies in Human/mouse Each vial provides a nominal titer of 1x10E6 transducing units TU miR-375-3p Lentivirus miRNA lentiviral particles for expression of human/mouse miR-375 (3p) The lentiviral vector contains the entire human miRNA stem-loop which leads to expression of the human/mouse common mature miRNA sequence in transduced cells The miRNA lentivirus is ultra-purified and concentrated by PEG precipitation and sucrose gradient centrifugation ideal for in vitro or in vivo use and transduction of difficult-to-transfect (e g thawed and/or primary) cells
Catalog No. 50-003-30746 Supplier BIOHIPPO Supplier No. BHV19400177
miR-375-3p-GFP-Puro lentiviral vector expressing microRNA 375 with GFP reporter and Puromycin selection for studies in Human/mouse Each vial provides a nominal titer of 1x10E6 transducing units TU miR-375-3p Lentivirus miRNA lentiviral particles for expression of human/mouse miR-375 (3p) The lentiviral vector contains the entire human miRNA stem-loop which leads to expression of the human/mouse common mature miRNA sequence in transduced cells The miRNA lentivirus is ultra-purified and concentrated by PEG precipitation and sucrose gradient centrifugation ideal for in vitro or in vivo use and transduction of difficult-to-transfect (e g thawed and/or primary) cells