Learn More
Perkin Elmer US LLC Polystyrene Test Film - MIR
Catalog No. 509043276
Encompass_Preferred
This reference material mounts in your FT-IR spectrometer, allowing you to quickly and easily check the mid infrared frequencies and overall accuracy of your FT-IR spectrometer.
Catalog No. 50-904-3276 Supplier Perkin Elmer US LLC Supplier No. L1202057
This reference material mounts in your FT-IR spectrometer, allowing you to quickly and easily check the mid infrared frequencies and overall accuracy of your FT-IR spectrometer.