Learn More
PHCbi MIR Series 4.3 cu.ft. Compact Microbiological Incubator
Catalog No. 502720050
Encompass_Preferred
The MIR-154-PA compact microbiological incubator is recognized as an exceptional unit suitable for a wide range of applications that require a temperature range of -10°C to +60°C. Programmable models include three-step functions useful for investigations involving microbiology, plant cell biology and more.Effective capacity: 4.34 cu.ft. (123 L)Temperature range: -10℃ to +60℃
Catalog No. 50-272-0050 Supplier PHCbi Supplier No. MIR154PA
Add to Cart
Add to Cart
Request a Quote
The MIR-154-PA compact microbiological incubator is recognized as an exceptional unit suitable for a wide range of applications that require a temperature range of -10°C to +60°C. Programmable models include three-step functions useful for investigations involving microbiology, plant cell biology and more.Effective capacity: 4.34 cu.ft. (123 L)Temperature range: -10℃ to +60℃