Learn More
PHCbi MIR Series 8.4 cu.ft. Microbiological Incubator
Catalog No. 502720049
Encompass_Preferred
The MIR-254-PA microbiological incubator is recognized as an exceptional unit suitable for a wide range of applications that require a temperature range of -10°C to +60°C. Programmable models include three-step functions useful for investigations involving microbiology, plant cell biology and more.The MIR-254-PA microbiological incubator provides an ergonomic cabinet design, versatile applications usage and a wide temperature range control.Effective capacity: 8.40 cu.ft. (238 L)Temperature range: -10℃ to +60℃
Catalog No. 50-272-0049 Supplier PHCbi Supplier No. MIR254PA
Add to Cart
Add to Cart
Request a Quote
The MIR-254-PA microbiological incubator is recognized as an exceptional unit suitable for a wide range of applications that require a temperature range of -10°C to +60°C. Programmable models include three-step functions useful for investigations involving microbiology, plant cell biology and more.The MIR-254-PA microbiological incubator provides an ergonomic cabinet design, versatile applications usage and a wide temperature range control.Effective capacity: 8.40 cu.ft. (238 L)Temperature range: -10℃ to +60℃